Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C13A2.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072446.4 | NP_504847.1 | 2 - 3 | 480 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |