Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028564.5 | NP_001023735.1 | 6 - 7 | 825 | 123 | |
NM_001028565.4 | NP_001023736.1 | 5 - 6 | 912 | 123 | ||
GenBank mRNA | AF497825.1 | - | 5 - 6 | 795 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() |