Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acd-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072829.3 | NP_505230.1 | 6 - 7 | 1184 | 109 |
Set Membership: |
![]() ![]() ![]() ![]() |