Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: add-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047665.4 | NP_001041130.1 | 6 - 7 | 1148 | 104 | |
NM_001047666.5 | NP_001041131.1 | 1 - 2 | 197 | 104 | ||
GenBank mRNA | AF166171.1 | - | 6 - 7 | 1148 | 104 |
Set Membership: |
![]() ![]() ![]() ![]() |