Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Y87G2A.19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026664.5 | NP_001021835.1 | 2 - 3 | 260 | 127 |
Set Membership: |
![]() ![]() ![]() ![]() |