Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C27F2.9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065652.6 | NP_498053.2 | 2 - 3 | 216 | 108 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |