Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C27F2.9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065652.6 | NP_498053.2 | 3 - 4 | 365 | 98 |
Gene Symbol: vps-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065651.7 | NP_498052.1 | 3 - 4 | 329 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |