Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R144.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379693.1 | NP_001367578.1 | 9 - 10 | 1070 | 116 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |