Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDKRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003095.1 | NP_001003095.1 | 1 - 2 | 137 | 65 | |
GenBank mRNA | AF334948.1 | - | 1 - 2 | 137 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() |