Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDKRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003095.1 | NP_001003095.1 | 2 - 3 | 251 | 59 | |
XM_005623175.3 | XP_005623232.1 | 3 - 4 | 832 | 59 | ||
GenBank mRNA | AF334948.1 | - | 2 - 3 | 251 | 59 |
Set Membership: |
![]() ![]() ![]() ![]() |