Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618088.3 | XP_005618145.1 | 470 | 131 | ||
XM_846466.5 | XP_851559.1 | 467 | 131 |
Set Membership: |
![]() ![]() ![]() ![]() |