Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Cf02690631_m1
          See other PTEN GE Assays ›
          Gene Symbol:
          PTEN
          Gene Name:
          phosphatase and tensin homolog
          Gene Aliases:
          MMAC1
          Chromosome Location:
          Chr.26: 37853148 - 37913176 on Build CanFam3.1
          Species:
          Dog
          Species Specific ID (Flybase ID):
          -
          Assay ID Cf02690631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTEN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001003192.1 NP_001003192.1 6 - 7 915 83
          GenBank mRNA U92435.1 - 6 - 7 915 83

          Back To Top

          More Information


          Additional Information:

          This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          regulation of B cell apoptotic process
          protein dephosphorylation
          apoptotic process
          neuron-neuron synaptic transmission
          central nervous system development
          heart development
          learning or memory
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          negative regulation of epithelial to mesenchymal transition
          regulation of neuron projection development
          cell migration
          dentate gyrus development
          central nervous system neuron axonogenesis
          negative regulation of cell migration
          adult behavior
          negative regulation of myelination
          regulation of protein stability
          regulation of synaptic transmission, GABAergic
          central nervous system myelin maintenance
          regulation of myeloid cell apoptotic process
          negative regulation of peptidyl-serine phosphorylation
          multicellular organismal response to stress
          social behavior
          peptidyl-tyrosine dephosphorylation
          maternal behavior
          negative regulation of apoptotic process
          protein kinase B signaling
          endothelial cell migration
          locomotor rhythm
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of cell size
          inositol phosphate dephosphorylation
          phosphatidylinositol dephosphorylation
          negative regulation of axon regeneration
          cardiac muscle tissue development
          forebrain morphogenesis
          negative regulation of epithelial cell proliferation
          negative regulation of axonogenesis
          protein stabilization
          positive regulation of DNA-binding transcription factor activity
          negative regulation of keratinocyte migration
          negative regulation of focal adhesion assembly
          negative regulation of protein kinase B signaling
          rhythmic synaptic transmission
          negative regulation of cardiac muscle cell proliferation
          canonical Wnt signaling pathway
          synapse maturation
          prepulse inhibition
          male mating behavior
          long-term synaptic potentiation
          prostate gland growth
          dendritic spine morphogenesis
          negative regulation of dendritic spine morphogenesis
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          cellular response to electrical stimulus
          cellular response to hypoxia
          negative regulation of ribosome biogenesis
          negative regulation of cell aging
          negative regulation of excitatory postsynaptic potential
          presynaptic membrane assembly
          postsynaptic density assembly
          negative regulation of wound healing, spreading of epidermal cells
          positive regulation of TRAIL-activated apoptotic signaling pathway
          positive regulation of ubiquitin protein ligase activity
          negative regulation of vascular associated smooth muscle cell proliferation
          positive regulation of ubiquitin-dependent protein catabolic process
          negative regulation of G1/S transition of mitotic cell cycle
          positive regulation of excitatory postsynaptic potential
          negative regulation of synaptic vesicle clustering
          phosphatidylinositol-3-phosphatase activity
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity
          protein tyrosine/serine/threonine phosphatase activity
          anaphase-promoting complex binding
          phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity
          protein kinase binding
          PDZ domain binding
          identical protein binding
          inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
          phosphatidylinositol-3,4-bisphosphate 3-phosphatase activity
          ubiquitin-specific protease binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0