Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Cf02711958_s1
See other LOC478038 GE Assays ›
Gene Symbol:
LOC478038
Gene Name:
protocadherin gamma-C5
Gene Aliases:
PCDHGA1, PCDHGA10, PCDHGA11, PCDHGA3, PCDHGA6, PCDHGA7, PCDHGB2, PCDHGB6, PCDHGB7, PCDHGC3
Chromosome Location:
Chr.2: 36297841 - 36481900 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID
Cf02711958_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: LOC478038
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_014107808.2
XP_013963283.1
2941
64
XM_014107813.2
XP_013963288.1
2767
64
XM_014107817.2
XP_013963292.1
2960
64
XM_014107820.2
XP_013963295.1
2961
64
XM_014107828.2
XP_013963303.1
3193
64
XM_014107832.2
XP_013963307.1
2956
64
XM_014107840.2
XP_013963315.1
2911
64
XM_014107851.2
XP_013963326.1
3159
64
XM_014107854.2
XP_013963329.1
2953
64
XM_014107859.2
XP_013963334.1
3190
64
XM_014107867.2
XP_013963342.1
3190
64
XM_014107871.2
XP_013963346.1
3044
64
XM_014107873.2
XP_013963348.1
2950
64
XM_014107874.2
XP_013963349.1
2942
64
XM_014107879.2
XP_013963354.2
2943
64
XM_014107883.2
XP_013963358.1
2981
64
XM_014107888.2
XP_013963363.1
2939
64
XM_014107889.2
XP_013963364.1
2943
64
XM_014107892.2
XP_013963367.1
2840
64
XM_014107899.2
XP_013963374.1
3441
64
XM_014107916.2
XP_013963391.2
381
64
XM_014107921.2
XP_013963396.1
311
64
XM_014107925.2
XP_013963400.1
557
64
XM_022408700.1
XP_022264408.1
2939
64
XM_022408729.1
XP_022264437.1
365
64
XM_022408734.1
XP_022264442.1
494
64
Back To Top
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership:
Amplicon length less than or equal to 70
3' Most
Canis familiaris
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0