Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197159.1 | NP_001184088.1 | 10 - 11 | 1517 | 69 | |
XM_005619468.3 | XP_005619525.1 | 10 - 11 | 1421 | 69 | ||
XM_005619469.3 | XP_005619526.1 | 10 - 11 | 1818 | 69 | ||
GenBank mRNA | AB512464.1 | - | 10 - 11 | 1518 | 69 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |