Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076602.1 | NP_001070070.1 | 6 - 7 | 788 | 92 | |
GenBank mRNA | BC124222.1 | - | 6 - 7 | 788 | 92 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |