Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cttnbp2nlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077371.1 | NP_001070839.1 | 2 - 3 | 228 | 64 | |
XM_005167035.4 | XP_005167092.1 | 2 - 3 | 239 | 64 | ||
GenBank mRNA | BC124670.1 | - | 2 - 3 | 228 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() |