Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pax9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131298.1 | NP_571373.1 | 2 - 3 | 778 | 87 | |
GenBank mRNA | U40931.1 | - | 2 - 3 | 778 | 87 |
Set Membership: |
![]() ![]() ![]() |