Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slit1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034968.1 | NP_001030140.1 | 27 - 28 | 2992 | 94 | |
XM_009307022.3 | XP_009305297.1 | 24 - 25 | 2711 | 94 | ||
GenBank mRNA | BC163538.1 | - | 27 - 28 | 2884 | 94 | |
DQ177286.1 | - | 27 - 28 | 2992 | 94 |
Set Membership: |
![]() ![]() ![]() ![]() |