Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pa2g4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212641.2 | NP_997806.1 | 3 - 4 | 389 | 72 | |
GenBank mRNA | BC056591.1 | - | 3 - 4 | 361 | 72 | |
BC071536.1 | - | 3 - 4 | 389 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |