Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: si:ch211-264f5.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098253.1 | NP_001091723.1 | 1 - 2 | 87 | 80 | |
GenBank mRNA | CU458917.1 | - | 1 - 2 | 87 | 80 |
Set Membership: |
![]() ![]() ![]() |