Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr6a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131256.2 | NP_571331.2 | 3 - 4 | 463 | 61 | |
XM_017357271.2 | XP_017212760.1 | 2 - 3 | 747 | 61 | ||
GenBank mRNA | AJ007703.2 | - | 3 - 4 | 463 | 61 | |
BC162651.1 | - | 2 - 3 | 304 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() |