Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk2a2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131240.1 | NP_571315.1 | 3 - 4 | 1199 | 136 | |
GenBank mRNA | BC044342.1 | - | 3 - 4 | 1182 | 136 | |
BC164160.1 | - | 3 - 4 | 349 | 136 | ||
X99964.1 | - | 1199 | 136 |
Set Membership: |
![]() ![]() ![]() ![]() |