Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045207.1 | NP_001038672.1 | 5 - 6 | 893 | 113 | |
XM_005158734.4 | XP_005158791.1 | 5 - 6 | 914 | 113 |
Set Membership: |
![]() ![]() ![]() |