Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03138226_mH
          See other PAX2A GE Assays ›
          Gene Symbol:
          pax2a
          Gene Name:
          paired box 2a
          Gene Aliases:
          PAXZF-B, Pax-2, cb378, noi, pax-b, pax2.1, pax2a1, pax[zf-b], paxb
          Chromosome Location:
          Chr.13: 29769732 - 29804925 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03138226_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: pax2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131184.2 NP_571259.1 1 - 2 547 61
          XM_005156735.4 XP_005156792.1 1 - 2 1768 61
          XM_005156744.4 XP_005156801.1 1 - 2 1190 61
          XM_005156747.4 XP_005156804.1 1 - 2 1183 61
          XM_005156748.4 XP_005156805.1 1 - 2 1184 61
          XM_017358679.2 XP_017214168.1 1 - 2 1773 61
          XM_017358683.2 XP_017214172.1 1 - 2 1765 61
          XM_017358684.2 XP_017214173.1 1 - 2 1280 61
          XM_017358685.2 XP_017214174.1 1 - 2 1201 61
          XM_021480513.1 XP_021336188.1 1 - 2 1183 61
          GenBank mRNA AF067530.1 - 1 - 2 43 61
          AF067531.1 - 1 - 2 43 61
          AF067533.1 - 1 - 2 43 61
          AF067535.1 - 1 - 2 43 61
          AF067536.1 - 1 - 2 43 61
          AF067539.1 - 1 - 2 43 61
          AF067540.1 - 1 - 2 43 61
          BC066690.1 - 1 - 2 547 61
          X63961.1 - 669 61

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate specification
          kidney development
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          multicellular organism development
          nervous system development
          peripheral nervous system development
          embryonic pattern specification
          anterior/posterior pattern specification
          cerebellum development
          cerebellum formation
          locus ceruleus development
          glial cell development
          cell differentiation
          thyroid gland development
          midbrain development
          hindbrain development
          midbrain-hindbrain boundary development
          anterior/posterior pattern specification involved in pronephros development
          pronephric glomerulus morphogenesis
          otic placode formation
          embryonic camera-type eye morphogenesis
          pronephros development
          anatomical structure development
          retina development in camera-type eye
          inner ear receptor cell differentiation
          vestibular receptor cell development
          cilium assembly
          pronephros morphogenesis
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0