Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03138230_m1
          See other PAX2A GE Assays ›
          Gene Symbol:
          pax2a
          Gene Name:
          paired box 2a
          Gene Aliases:
          PAXZF-B, Pax-2, cb378, noi, pax-b, pax2.1, pax2a1, pax[zf-b], paxb
          Chromosome Location:
          Chr.13: 29769732 - 29804925 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03138230_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: pax2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131184.2 NP_571259.1 8 - 9 1522 62
          XM_005156735.4 XP_005156792.1 10 - 11 3049 62
          XM_005156744.4 XP_005156801.1 9 - 10 2210 62
          XM_005156747.4 XP_005156804.1 9 - 10 2203 62
          XM_005156748.4 XP_005156805.1 8 - 9 2159 62
          XM_017358679.2 XP_017214168.1 11 - 12 3075 62
          XM_017358683.2 XP_017214172.1 10 - 11 2995 62
          XM_017358684.2 XP_017214173.1 9 - 10 2465 62
          XM_017358685.2 XP_017214174.1 9 - 10 2386 62
          XM_021480493.1 XP_021336168.1 11 - 12 3088 62
          XM_021480494.1 XP_021336169.1 10 - 11 3061 62
          XM_021480495.1 XP_021336170.1 11 - 12 3085 62
          XM_021480496.1 XP_021336171.1 10 - 11 3041 62
          XM_021480497.1 XP_021336172.1 11 - 12 3032 62
          XM_021480498.1 XP_021336173.1 10 - 11 3007 62
          XM_021480499.1 XP_021336174.1 10 - 11 2986 62
          XM_021480500.1 XP_021336175.1 9 - 10 2480 62
          XM_021480501.1 XP_021336176.1 9 - 10 2400 62
          XM_021480502.1 XP_021336177.1 10 - 11 2871 62
          XM_021480503.1 XP_021336178.1 10 - 11 2869 62
          XM_021480504.1 XP_021336179.1 10 - 11 2330 62
          XM_021480505.1 XP_021336180.1 9 - 10 2223 62
          XM_021480506.1 XP_021336181.1 9 - 10 2208 62
          XM_021480507.1 XP_021336182.1 9 - 10 2216 62
          XM_021480508.1 XP_021336183.1 8 - 9 2172 62
          XM_021480509.1 XP_021336184.1 8 - 9 2165 62
          XM_021480510.1 XP_021336185.1 7 - 8 2533 62
          XM_021480511.1 XP_021336186.1 7 - 8 2477 62
          XM_021480512.1 XP_021336187.1 6 - 7 1874 62
          XM_021480513.1 XP_021336188.1 6 - 7 1861 62
          GenBank mRNA AF067530.1 - 9 - 10 1063 62
          AF067531.1 - 8 - 9 1018 62
          AF067532.1 - 9 - 10 1141 62
          AF067533.1 - 7 - 8 898 62
          AF067534.1 - 9 - 10 1075 62
          AF067535.1 - 6 - 7 721 62
          AF067536.1 - 9 - 10 1129 62
          AF067537.1 - 9 - 10 1075 62
          AF067538.1 - 8 - 9 1030 62
          AF067539.1 - 8 - 9 1018 62
          AF067540.1 - 9 - 10 1063 62
          AF067541.1 - 8 - 9 1030 62
          BC066690.1 - 8 - 9 1522 62
          X63961.1 - 1644 62

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate specification
          kidney development
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          multicellular organism development
          nervous system development
          peripheral nervous system development
          embryonic pattern specification
          anterior/posterior pattern specification
          cerebellum development
          cerebellum formation
          locus ceruleus development
          glial cell development
          cell differentiation
          thyroid gland development
          midbrain development
          hindbrain development
          midbrain-hindbrain boundary development
          anterior/posterior pattern specification involved in pronephros development
          pronephric glomerulus morphogenesis
          otic placode formation
          embryonic camera-type eye morphogenesis
          pronephros development
          anatomical structure development
          retina development in camera-type eye
          inner ear receptor cell differentiation
          vestibular receptor cell development
          cilium assembly
          pronephros morphogenesis
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0