Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030178.2 | NP_001025349.2 | 17 - 18 | 4971 | 74 | |
XM_005172621.4 | XP_005172678.1 | 17 - 18 | 5026 | 74 | ||
XM_005172623.4 | XP_005172680.1 | 17 - 18 | 5022 | 74 | ||
XM_021479186.1 | XP_021334861.1 | 17 - 18 | 4934 | 74 |
Set Membership: |
![]() ![]() ![]() |