Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmbox1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030264.1 | NP_001025435.1 | 6 - 7 | 1183 | 65 | |
XM_005169831.4 | XP_005169888.1 | 6 - 7 | 1098 | 65 | ||
XM_005169832.4 | XP_005169889.1 | 6 - 7 | 1291 | 65 | ||
XM_009292894.3 | XP_009291169.1 | 6 - 7 | 1296 | 65 | ||
XM_021467220.1 | XP_021322895.1 | 6 - 7 | 1295 | 65 | ||
XM_021467221.1 | XP_021322896.1 | 6 - 7 | 1292 | 65 | ||
GenBank mRNA | BC097126.1 | - | 6 - 7 | 1183 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |