Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: actn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131522.2 | NP_571597.1 | 2 - 3 | 398 | 79 | |
GenBank mRNA | BC054911.1 | - | 2 - 3 | 399 | 79 | |
FJ979801.1 | - | 2 - 3 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() |