Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: f7i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173228.1 | NP_775335.1 | 4 - 5 | 391 | 68 | |
GenBank mRNA | AF515269.1 | - | 4 - 5 | 391 | 68 | |
BC094994.1 | - | 4 - 5 | 369 | 68 | ||
BC115091.1 | - | 4 - 5 | 375 | 68 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |