Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calhm5.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089465.2 | NP_001082934.2 | 2 | 1569 | 147 | |
GenBank mRNA | BC115341.1 | - | 1553 | 148 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |