Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lpar5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199606.1 | NP_955900.1 | 1 | 1449 | 131 | |
GenBank mRNA | BC056587.1 | - | 1 | 1449 | 131 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |