Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacnb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113519.2 | NP_001106991.1 | 8 - 9 | 814 | 99 | |
XM_005167837.4 | XP_005167894.1 | 7 - 8 | 882 | 99 | ||
GenBank mRNA | BC171593.1 | - | 8 - 9 | 720 | 99 | |
BC171597.1 | - | 8 - 9 | 720 | 99 | ||
DQ372948.1 | - | 7 - 8 | 690 | 99 | ||
EF601060.1 | - | 7 - 8 | 827 | 99 | ||
EF601061.1 | - | 8 - 9 | 818 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() |