Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03436992_m1
          See other CLASP1A GE Assays ›
          Gene Symbol:
          clasp1a
          Gene Name:
          cytoplasmic linker associated protein 1a
          Gene Aliases:
          si:dkey-219c10.5
          Chromosome Location:
          Chr.9: 38163853 - 38291813 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03436992_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: clasp1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001115139.1 NP_001108611.1 39 - 40 4519 81
          XM_009304822.3 XP_009303097.1 42 - 43 5083 81
          XM_009304834.3 XP_009303109.1 39 - 40 4834 81
          XM_009304836.3 XP_009303111.1 38 - 39 4807 81
          XM_009304838.3 XP_009303113.1 37 - 38 4783 81
          XM_017357687.2 XP_017213176.1 43 - 44 5180 81
          XM_021478695.1 XP_021334370.1 44 - 45 5321 81
          XM_021478696.1 XP_021334371.1 44 - 45 5318 81
          XM_021478697.1 XP_021334372.1 44 - 45 5306 81
          XM_021478698.1 XP_021334373.1 43 - 44 5297 81
          XM_021478699.1 XP_021334374.1 43 - 44 5297 81
          XM_021478700.1 XP_021334375.1 43 - 44 5294 81
          XM_021478701.1 XP_021334376.1 42 - 43 5270 81
          XM_021478702.1 XP_021334377.1 44 - 45 5267 81
          XM_021478703.1 XP_021334378.1 43 - 44 5258 81
          XM_021478704.1 XP_021334379.1 43 - 44 5249 81
          XM_021478705.1 XP_021334380.1 42 - 43 5234 81
          XM_021478706.1 XP_021334381.1 43 - 44 5225 81
          XM_021478707.1 XP_021334382.1 43 - 44 5204 81
          XM_021478708.1 XP_021334383.1 42 - 43 5147 81
          XM_021478709.1 XP_021334384.1 42 - 43 5096 81
          XM_021478710.1 XP_021334385.1 41 - 42 5072 81
          XM_021478711.1 XP_021334386.1 40 - 41 5059 81
          XM_021478712.1 XP_021334387.1 41 - 42 4999 81
          XM_021478713.1 XP_021334388.1 40 - 41 4975 81
          XM_021478714.1 XP_021334389.1 40 - 41 4972 81
          XM_021478715.1 XP_021334390.1 39 - 40 4948 81
          XM_021478716.1 XP_021334391.1 39 - 40 4948 81
          XM_021478717.1 XP_021334392.1 38 - 39 4924 81
          XM_021478718.1 XP_021334393.1 37 - 38 4261 81

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          regulation of microtubule polymerization or depolymerization
          establishment of mitotic spindle localization
          mitotic spindle assembly
          microtubule binding
          kinetochore binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c8579b687-54cxl:80/100.66.135.11:80.
          git-commit: 8e87205ac1279514f9c7a0d34bcd4a43061862b7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0