Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916356.4 | XP_001916391.1 | 273 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() |