Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205195.1 | NP_990526.1 | 1 | 949 | 82 | |
GenBank mRNA | X98283.1 | - | 949 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |