Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021082.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021082.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021082.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021082.hg.1 |
TC01001346.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021081.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021081.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021081.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021081.hg.1 |
TC01001346.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175105.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175105.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175105.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175105.hg.1 |
TC0100010241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175105.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175105.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175105.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000438394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010241.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175107.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175107.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175107.hg.1 |
NM_002432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175107.hg.1 |
TC0100010241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175107.hg.1 |
ENST00000368141.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175107.hg.1 |
ENST00000491210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175107.hg.1 |
NM_002432.2 |
Refseq |