Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019473.hg.1 |
ENST00000370311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019473.hg.1 |
ENST00000370314.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019473.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019473.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019473.hg.1 |
TC0X001493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000417858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000417858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000497894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000497894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019476.hg.1 |
ENST00000370314.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019476.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019476.hg.1 |
TC0X001493.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0X00175415.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175415.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175415.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175415.hg.1 |
TC0X00011126.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175415.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175415.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175415.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175416.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175416.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175416.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175416.hg.1 |
TC0X00011126.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175416.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175416.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175416.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000417858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000417858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000497894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000497894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
ENST00000535043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011126.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175418.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175418.hg.1 |
NM_000808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175418.hg.1 |
TC0X00011126.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175418.hg.1 |
ENST00000370314.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175418.hg.1 |
NM_000808.3 |
Refseq |