Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00171054_m1
          See other CCR7 GE Assays ›
          Gene Symbol:
          CCR7
          Gene Name:
          C-C motif chemokine receptor 7
          Gene Aliases:
          BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1
          Chromosome Location:
          Chr.17: 40553769 - 40565484 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00171054_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCR7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001838.3 NP_001829.1 1 - 2 84 64
          GenBank mRNA AK127810.1 - 1 - 2 72 64
          AK312732.1 - 1 - 2 68 64
          BC035343.1 - 1 - 2 50 5180758 64
          DA814913.1 - 1 - 2 84 64
          L08176.1 - 1 - 2 72 64
          L31581.1 - 1 - 2 75 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010630.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001466.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK127810.1,AK312732.1,BC035343.1,L08176.1,L31581.1,NM_001838.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Immune Response 1700 Inventoried MicroArray Quality Control (MAQC) Human_Immune Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCR7

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of T cell polarity
          positive regulation of cell-matrix adhesion
          dendritic cell chemotaxis
          myeloid dendritic cell chemotaxis
          positive regulation of dendritic cell antigen processing and presentation
          positive regulation of hypersensitivity
          positive regulation of humoral immune response
          inflammatory response
          immune response
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          positive regulation of actin filament polymerization
          positive regulation of pseudopodium assembly
          ruffle organization
          response to lipopolysaccharide
          regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          response to prostaglandin E
          chemokine (C-C motif) ligand 19 signaling pathway
          chemokine (C-C motif) ligand 21 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of phosphatidylinositol 3-kinase activity
          negative thymic T cell selection
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          homeostasis of number of cells
          regulation of interleukin-1 beta secretion
          positive regulation of T cell receptor signaling pathway
          release of sequestered calcium ion into cytosol
          positive regulation of filopodium assembly
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cytokine stimulus
          response to nitric oxide
          interleukin-12 secretion
          positive regulation of neutrophil chemotaxis
          activation of GTPase activity
          lymphocyte migration into lymph node
          mature conventional dendritic cell differentiation
          negative regulation of leukocyte apoptotic process
          positive regulation of cell motility
          positive regulation of dendritic cell chemotaxis
          positive regulation of immunological synapse formation
          positive regulation of T cell costimulation
          positive regulation of glycoprotein biosynthetic process involved in immunological synapse formation
          regulation of dendritic cell dendrite assembly
          G-protein coupled receptor activity
          C-C chemokine receptor activity
          chemokine (C-C motif) ligand 19 binding
          chemokine (C-C motif) ligand 21 binding
          C-C motif chemokine 19 receptor activity
          C-C motif chemokine 21 receptor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000246657.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000246657.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000246657.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000246657.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000246657.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000246657.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000578085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000578085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000579344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000579344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000579344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000579344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000579344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 ENST00000579344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301717.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301717.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301717.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301717.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301717.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301717.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001301718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001838.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001838.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001838.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001838.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001838.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010630.hg.1 NM_001838.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 186454 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix