Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00184839_m1
          See other BCL10 GE Assays ›
          Gene Symbol:
          BCL10
          Gene Name:
          B-cell CLL/lymphoma 10
          Gene Aliases:
          CARMEN, CIPER, CLAP, IMD37, c-E10, mE10
          Chromosome Location:
          Chr.1: 85265776 - 85276904 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00184839_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BCL10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320715.1 NP_001307644.1 1 - 2 611 146
          NM_003921.4 NP_003912.1 1 - 2 611 146
          GenBank mRNA AF057700.1 - 1 - 2 305 146
          AF082283.1 - 2 - 3 765 146
          AF100338.1 - 1 - 2 306 146
          AF134395.1 - 1 - 2 304 146
          AJ006288.1 - 2 - 3 749 146
          AK291346.1 - 1 - 2 269 146
          BC053617.1 - 1 - 2 181 6143423 146
          BX647306.1 - 1 - 2 224 146
          EF189176.1 - 1 - 2 164 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014775.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002828.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF057700.1, AF082283.1, AF100338.1, AF134395.1, AJ006288.1, AK291346.1, BC053617.1, EF189176.1, NM_001320715.1, NM_003921.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human NFkB Pathway 1700 Inventoried Human_Nfkb Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BCL10

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell apoptotic process
          neural tube closure
          stimulatory C-type lectin receptor signaling pathway
          toll-like receptor signaling pathway
          response to molecule of bacterial origin
          adaptive immune response
          negative regulation of mature B cell apoptotic process
          cellular defense response
          I-kappaB kinase/NF-kappaB signaling
          cell death
          response to fungus
          immunoglobulin mediated immune response
          positive regulation of protein ubiquitination
          response to food
          lymphotoxin A production
          positive regulation of mast cell cytokine production
          Fc-epsilon receptor signaling pathway
          positive regulation of phosphorylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          innate immune response
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          regulation of T cell receptor signaling pathway
          positive regulation of T cell activation
          positive regulation of NF-kappaB transcription factor activity
          protein complex oligomerization
          protein homooligomerization
          protein heterooligomerization
          T cell apoptotic process
          cellular response to mechanical stimulus
          positive regulation of extrinsic apoptotic signaling pathway
          protease binding
          transcription coactivator activity
          protein binding
          protein C-terminus binding
          transcription factor binding
          kinase activator activity
          enzyme binding
          kinase binding
          protein kinase binding
          ubiquitin protein ligase binding
          identical protein binding
          ubiquitin binding
          protein kinase B binding
          protein self-association
          NF-kappaB binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01044015.hg.1 ENST00000370580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044015.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01044015.hg.1 TC01002828.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01044011.hg.1 ENST00000271015.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044011.hg.1 ENST00000370580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044011.hg.1 ENST00000394761.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044011.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01044011.hg.1 TC01002828.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01044012.hg.1 ENST00000271015.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044012.hg.1 ENST00000370580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044012.hg.1 ENST00000394761.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044012.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01044012.hg.1 TC01002828.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01044014.hg.1 ENST00000271015.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044014.hg.1 ENST00000370580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044014.hg.1 NM_003921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01044014.hg.1 TC01002828.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100210075.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210075.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210075.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210075.hg.1 TC0100014775.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100210075.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210075.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210075.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210079.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210079.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210079.hg.1 TC0100014775.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100210079.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210079.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay TC0100014775.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210076.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210076.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210076.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210076.hg.1 TC0100014775.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100210076.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210076.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210076.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210077.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210077.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210077.hg.1 NM_003921.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100210077.hg.1 TC0100014775.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100210077.hg.1 ENST00000370580.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210077.hg.1 ENST00000620248.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210077.hg.1 NM_003921.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 195465 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0