Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00194498_m1
          See other REST GE Assays ›
          Gene Symbol:
          REST
          Gene Name:
          RE1 silencing transcription factor
          Gene Aliases:
          NRSF, WT6, XBR
          Chromosome Location:
          Chr.4: 56907876 - 56935847 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00194498_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: REST
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005612.4 NP_005603.3 1 - 2 337 79
          XM_017008527.1 XP_016864016.1 1 - 2 298 79
          GenBank mRNA BC038985.1 - 1 - 2 266 79
          JX896957.1 - 1 - 2 119 79
          JX896958.1 - 1 - 2 119 79
          JX896959.1 - 1 - 2 119 79
          JX896971.1 - 1 - 2 119 79
          JX896978.1 - 1 - 2 119 79
          JX896979.1 - 1 - 2 119 79
          JX896983.1 - 1 - 2 119 79
          JX896984.1 - 1 - 2 119 79
          JX896991.1 - 1 - 2 119 79
          KC117266.1 - 1 - 2 119 79
          U13879.1 - 1 - 2 237 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400007609.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000335.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC038985.1,JX896957.1,JX896958.1,JX896959.1,JX896971.1,JX896978.1,JX896979.1,JX896983.1,JX896984.1,JX896991.1,KC117266.1,NM_005612.4,XM_017008527.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human Stem Cell Pluripotency Human Transcriptional Regulatory Network in Embryonic Stem C HUMAN HUNTINGTON DISEASE PATHWAY Inventoried Human_Stem_Cell Human_Transcriptional_Regulatory_Network_in_Embryonic_Stem_Cell Human_Huntington_Disease_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene REST

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          hematopoietic progenitor cell differentiation
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          negative regulation of cell proliferation
          negative regulation of gene expression
          negative regulation of aldosterone biosynthetic process
          cellular response to drug
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation by host of viral transcription
          negative regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of calcium ion-dependent exocytosis
          negative regulation of insulin secretion
          negative regulation of neurogenesis
          cardiac muscle cell myoblast differentiation
          histone H4 deacetylation
          cellular response to electrical stimulus
          cellular response to glucocorticoid stimulus
          potassium ion transmembrane transport
          negative regulation of cortisol biosynthetic process
          negative regulation of dense core granule biogenesis
          negative regulation of mesenchymal stem cell differentiation
          negative regulation of amniotic stem cell differentiation
          core promoter sequence-specific DNA binding
          core promoter binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          outward rectifier potassium channel activity
          protein complex binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000309042.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000503522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000503522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000511065.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000511065.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000511065.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000511065.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000514063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000514063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000514063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000514063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000514063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000514063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000611211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000612429.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000616975.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000619101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000622863.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000622863.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000622863.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000622863.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000622863.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000622863.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 ENST00000622863.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_001193508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007609.hg.1 NM_005612.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0