Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000219097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000563599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000566860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000567000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000568364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000569239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000569239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000569239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000569239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000569239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000569239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000570260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000570260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000570260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000570260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000570260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000570260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
ENST00000575571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NM_014321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007718.hg.1 |
NR_037620.1 |
Refseq |