Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00253644_s1
          See other PRM3 GE Assays ›
          Gene Symbol:
          PRM3
          Gene Name:
          protamine 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 11273199 - 11273595 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00253644_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRM3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021247.2 NP_067070.2 1 240 70
          GenBank mRNA AW666005.1 - 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRM3

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          biological_process
          cell differentiation
          chromosome condensation
          flagellated sperm motility
          molecular_function
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30439205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09927223 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3363732 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P305470 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11413 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58902 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3363732 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3363732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30439205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30439205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09927224 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P305470 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927238 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927228 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927226 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927236 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927241 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927232 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927233 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09927223 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P305470 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927178 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927253 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16851396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927203 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927263 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927232 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927251 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927256 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927226 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927259 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927258 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927233 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927176 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16851381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927255 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927236 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927197 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927261 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927241 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16851379 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927267 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927249 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927196 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927223 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927209 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927238 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927228 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927195 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927190 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927244 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927265 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16851390 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09927207 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_021247_at - -
          Affymetrix Human Gene 1.0 ST Array 7999432 - -
          Affymetrix Human Gene 1.1 ST Array 7999431 - -
          Affymetrix HT HG-U133+ PM Array Plate 231758_PM_at - -
          Affymetrix Human Genome U219 Array 11743217_at - -
          Affymetrix Human Genome U219 Array 11743217_PM_at - -
          Human Genome U133 Plus 2.0 Array 231758_at - -
          Human Genome U133 Plus 2.0 Array 231758_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_021247_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231758_at4 - -
          Human Genome U133 Set 231758_PM_at - -
          Human Genome U133 Set 231758_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053483 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053483 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 178226 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1652284 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1667101 NM_021247.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1745027 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1789482 NM_021247.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1667101 NM_021247.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_10864056-S NM_021247.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_16955-A - -
          Illumina human-6 v1.0 expression beadchip 0002650068 - -
          Illumina humanRef-8 v1.0 expression beadchip 0002650068 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1667101 NM_021247.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1745027 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1652284 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1667101 NM_021247.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1820382 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_16955 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1789482 NM_021247.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1667101 NM_021247.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1667101 NM_021247.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1652284 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1789482 NM_021247.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1789482 NM_021247.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1667101 NM_021247.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652284 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1789482 NM_021247.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1667101 NM_021247.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix