Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00271467_m1
          See other IFI27 GE Assays ›
          Gene Symbol:
          IFI27
          Gene Name:
          interferon alpha inducible protein 27
          Gene Aliases:
          FAM14D, ISG12, ISG12A, P27
          Chromosome Location:
          Chr.14: 94110733 - 94116699 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00271467_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IFI27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130080.2 NP_001123552.1 2 - 3 219 63
          NM_001288952.1 NP_001275881.1 3 - 4 272 63
          NM_001288954.1 NP_001275883.1 3 - 4 272 63
          NM_001288956.1 NP_001275885.1 2 - 3 185 63
          NM_001288957.1 NP_001275886.1 2 - 3 284 63
          NM_001288958.1 NP_001275887.1 2 - 3 149 63
          NM_001288995.1 NP_001275924.1 2 - 3 185 63
          NM_005532.4 NP_005523.3 2 - 3 219 63
          GenBank mRNA AJ294851.2 - 2 - 3 180 63
          AK289535.1 - 2 - 3 217 63
          AK312089.1 - 2 - 3 216 63
          BC015492.1 - 2 - 3 284 3915505 63
          BF971643.1 - 3 - 4 251 63
          BG483267.1 - 2 - 3 183 63
          BM841506.1 - 2 - 3 182 63
          BM841596.1 - 2 - 3 216 63
          BP317624.1 - 3 - 4 269 63
          BT006781.1 - 1 - 2 96 63
          BU601451.1 - 2 - 3 219 63
          BU688913.1 - 3 - 4 63
          BX647330.1 - 2 - 3 216 63
          X67325.1 - 1 - 2 150 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008056.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000584.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IFI27

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of protein export from nucleus
          type I interferon signaling pathway
          apoptotic signaling pathway
          RNA polymerase II activating transcription factor binding
          molecular_function
          lamin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000555819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000557700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000557700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000557700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000557700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000611954.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000611954.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000611954.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000612813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614288.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614288.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614288.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614288.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614288.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614288.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000614648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000616764.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618200.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618200.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618200.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618200.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618200.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000618863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000620396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 ENST00000621160.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_001288995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008056.hg.1 NM_005532.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0