Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00277069_m1
          See other IRF4 GE Assays ›
          Gene Symbol:
          IRF4
          Gene Name:
          interferon regulatory factor 4
          Gene Aliases:
          LSIRF, MUM1, NF-EM5, SHEP8
          Chromosome Location:
          Chr.6: 391739 - 411443 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00277069_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IRF4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002460.3 NP_002451.2 4 - 5 620 105
          XM_006715090.2 XP_006715153.1 4 - 5 607 105
          GenBank mRNA AK310149.1 - 4 - 5 607 105
          BC015752.1 - 4 - 5 567 4861223 105
          U63738.1 - 4 - 5 599 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006442.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000005.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IRF4

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription by RNA polymerase II
          peptidyl-lysine methylation
          negative regulation of toll-like receptor signaling pathway
          T cell activation
          defense response to protozoan
          myeloid dendritic cell differentiation
          positive regulation of DNA binding
          histone H3 acetylation
          histone H4 acetylation
          regulation of T-helper cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          T-helper 17 cell lineage commitment
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity
          protein binding
          transcription factor binding
          protein-lysine N-methyltransferase activity
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06000041.hg.1 ENST00000380956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000041.hg.1 ENST00000412334.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000041.hg.1 ENST00000495137.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000041.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000041.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000041.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000041.hg.1 TC06000005.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 NR_046000.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 TC0600006442.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 NM_002460.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600144123.hg.1 NR_046000.2 Refseq

          Back To Top
           

          Citations (1)

          Role of the long cytoplasmic domain of the SIV Env glycoprotein in early and late stages of infection.
          Authors
          Vzorov AN, Weidmann A, Kozyr NL, Khaoustov V, Yoffe B, Compans RW
          Journal
          Retrovirology - Page No 94 (Mon Jan 01 00:00:00 UTC 2007)
          Volume (Year)
          4 (2007)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0