Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020852.hg.1 |
TC19001530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020852.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020852.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010693.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188262.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188262.hg.1 |
ENST00000593586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188262.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188262.hg.1 |
TC1900010693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188262.hg.1 |
NM_152479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188262.hg.1 |
ENST00000311308.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188262.hg.1 |
ENST00000593586.1 |
Ensembl |