Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00292955_s1
          See other FOXD2-AS1 GE Assays ›
          Gene Symbol:
          FOXD2-AS1
          Gene Name:
          FOXD2 antisense RNA 1 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 47432133 - 47434641 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00292955_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FOXD2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026878.1 1 352 84
          GenBank mRNA BC006113.1 - 1 352 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014090.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 122530 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P523096 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P523096 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 28595 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28983 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P523096 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P523096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181964 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181989 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181984 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181961 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181975 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15057833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181961 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15057833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181975 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181984 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181989 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181964 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20208039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00181996 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P523096 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P523096 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00059606 - -
          Agilent Human 1B Microarray G4111A A_32_P523096 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182018 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181957 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057827 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057878 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057835 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182008 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181959 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181964 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181999 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182003 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181945 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181930 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181984 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181941 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181993 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057883 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057853 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181932 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181939 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182038 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182019 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182026 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057870 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181986 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181972 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182002 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181948 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182010 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057864 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181936 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057881 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181927 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181943 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181929 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057839 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057859 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181934 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182009 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181968 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181996 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182015 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181969 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181980 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182032 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181975 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181976 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182034 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057877 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181989 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182020 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057854 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181951 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057843 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182037 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181938 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057862 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00181995 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15057845 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182022 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182012 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00182007 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 224456_PM_s_at - -
          Affymetrix HT HG-U133+ PM Array Plate 224457_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100014090.hg.1 ENST00000445551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014090.hg.1 ENST00000445551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014090.hg.1 NR_026878.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014090.hg.1 NR_026878.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100205520.hg.1 ENST00000445551.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205520.hg.1 NR_026878.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100205520.hg.1 TC0100014090.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100205520.hg.1 NR_026878.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100205520.hg.1 ENST00000445551.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014090.hg.1 ENST00000445551.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014090.hg.1 ENST00000445551.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014090.hg.1 NR_026878.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014090.hg.1 NR_026878.1 Refseq
          Human Genome U133 Plus 2.0 Array 224456_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 224456_s_at - -
          Human Genome U133 Plus 2.0 Array 224457_PM_at - -
          Human Genome U133 Plus 2.0 Array 224457_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224456_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224457_at5 - -
          Human Genome U95 Set 74159_PM_at - -
          Human Genome U95 Set 74159_at - -
          Human Genome U133 Set 224456_s_at - -
          Human Genome U133 Set 224457_PM_at - -
          Human Genome U133 Set 224457_at - -
          Human Genome U133 Set 224456_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045288 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045288 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309468 BC022099.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3309468 BC041637.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3309468 NR_026878.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3309468 BC006113.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309468 BC022099.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309468 BC041637.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309468 NR_026878.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309468 BC006113.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309468 BC022099.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309468 BC041637.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309468 NR_026878.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309468 BC006113.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309468 BC022099.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309468 BC041637.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309468 NR_026878.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309468 BC006113.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1852522 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 6370193 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_76479 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1852522 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1852522 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1852522 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix