Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00362799_s1
          See other FAM215A GE Assays ›
          Gene Symbol:
          FAM215A
          Gene Name:
          family with sequence similarity 215 member A (non-protein coding)
          Gene Aliases:
          APR-2, C17orf88, LINC00530
          Chromosome Location:
          Chr.17: 43917208 - 43917987 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00362799_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FAM215A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026770.1 1 255 102
          GenBank mRNA AF143236.1 - 1 267 102
          BC069097.1 - 1 255 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream membrane palmitoylated protein 2 MPP2 DLG2 -7.5kb Anti-sense Chr. 17 - 43875357 - 43909711 on NCBI Build GRCh38
          Downstream Wnt family member 9-like LOC102724183 11.9kb Sense Chr. 17 - 43927595 - 43934517 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007971.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000560.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10332344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10332352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10332346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10332348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16985111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16985107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30905638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10332349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10332345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10332351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16985108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16985086 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3387566 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P22562 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43995 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18650 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3387566 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00802260 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3387566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332321 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16985049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332322 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905600 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332321 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16985049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905600 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30905594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332322 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10332325 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P22562 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 5102 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 5211 - -
          Agilent Human 2 cDNA Microarray (G4101A) 3807982 - -
          Agilent Human 2 cDNA Microarray (G4101A) 3807982 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16985046 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332291 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332297 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332296 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332331 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332338 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332321 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332319 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332327 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332304 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332351 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332293 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332335 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332329 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332314 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16985112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332299 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332333 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332317 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332300 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332352 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332292 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10332310 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8007549 - -
          Affymetrix Human Gene 1.1 ST Array 8007548 - -
          Affymetrix HT HG-U133+ PM Array Plate 221129_PM_at - -
          Affymetrix Human Genome U219 Array 11750532_PM_x_at - -
          Affymetrix Human Genome U219 Array 11750532_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR17006816.hg.1 NR_026770.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006816.hg.1 TC17000560.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007971.hg.1 ENST00000588043.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007971.hg.1 ENST00000632077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007971.hg.1 NR_026770.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700158254.hg.1 ENST00000588043.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158254.hg.1 ENST00000632077.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158254.hg.1 NR_026770.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700158254.hg.1 TC1700007971.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700158254.hg.1 NR_026770.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700158254.hg.1 ENST00000588043.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158254.hg.1 ENST00000632077.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012417.hg.1 ENST00000591374.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007971.hg.1 ENST00000588043.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007971.hg.1 ENST00000632077.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007971.hg.1 NR_026770.1 Refseq
          Human Genome U133 Plus 2.0 Array 221129_at - -
          Human Genome U133 Plus 2.0 Array 221129_PM_at - -
          Human Genome U133A 2.0 Array 221129_at - -
          Human Genome U133A 2.0 Array 221129_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw550819 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221129_at8 - -
          Human Genome U133 Set 221129_at - -
          Human Genome U133 Set 221129_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009567 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009567 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3263062 NR_026770.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3263062 AF143236.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3263062 BC069097.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3263062 DA188505.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3194613 NR_026770.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3194613 AF143236.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3194613 BC069097.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline