Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00375457_m1
          See other P2RY12 GE Assays ›
          Gene Symbol:
          P2RY12
          Gene Name:
          purinergic receptor P2Y12
          Gene Aliases:
          ADPG-R, BDPLT8, HORK3, P2T(AC), P2Y(12)R, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12, SP1999
          Chromosome Location:
          Chr.3: 151336843 - 151384812 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00375457_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2RY12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022788.4 NP_073625.1 1 - 2 118 87
          GenBank mRNA AK292096.1 - 1 - 2 57 87
          AK303714.1 - 1 - 2 59 87
          AK316575.1 - 1 - 2 60 87
          BC017898.1 - 1 - 2 62 4251263 87
          DC313627.1 - 1 - 2 118 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300012816.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001907.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK292096.1, AK303714.1, AK316575.1, BC017898.1, NM_022788.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2RY12

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled adenosine receptor signaling pathway
          substrate-dependent cell migration, cell extension
          G protein-coupled receptor signaling pathway
          adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
          hemostasis
          glial cell migration
          negative regulation of norepinephrine secretion
          G protein-coupled purinergic nucleotide receptor signaling pathway
          positive regulation of ion transport
          protein kinase B signaling
          positive regulation of GTPase activity
          negative regulation of cell differentiation
          regulation of calcium ion transport
          platelet aggregation
          calcium ion transmembrane transport
          cellular response to ATP
          potassium ion transmembrane transport
          regulation of microglial cell migration
          G protein-coupled adenosine receptor activity
          G protein-coupled ADP receptor activity
          guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034369.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034369.hg.1 ENST00000455408.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034369.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034369.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034369.hg.1 TC03001907.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03034372.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034372.hg.1 ENST00000455408.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034372.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034372.hg.1 TC03001907.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 TC0300012816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300195929.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300195933.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195933.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300195933.hg.1 TC0300012816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300195933.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195933.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300195932.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195932.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300195932.hg.1 TC0300012816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300195932.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195932.hg.1 NM_022788.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 123625 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc