Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037333.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037333.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037333.hg.1 |
TC02005043.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200204825.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204825.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204825.hg.1 |
TC0200014382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200204825.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204825.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000281924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000467316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
ENST00000476823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014382.hg.1 |
NM_030923.4 |
Refseq |