Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00381264_m1
          See other ACBD5 GE Assays ›
          Gene Symbol:
          ACBD5
          Gene Name:
          acyl-CoA binding domain containing 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 27182624 - 27248674 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00381264_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACBD5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042473.3 NP_001035938.1 8 - 9 1003 123
          NM_001271512.3 NP_001258441.1 7 - 8 1317 123
          NM_001301251.1 NP_001288180.1 7 - 8 882 123
          NM_001301252.1 NP_001288181.1 6 - 7 1196 123
          NM_001301253.1 NP_001288182.1 7 - 8 872 123
          NM_001301254.1 NP_001288183.1 5 - 6 992 123
          NM_145698.4 NP_663736.2 8 - 9 1128 123
          XM_006717528.3 XP_006717591.3 9 - 10 7561 123
          XM_006717530.3 XP_006717593.1 9 - 10 1160 123
          XM_006717531.2 XP_006717594.1 9 - 10 1170 123
          XM_006717535.2 XP_006717598.1 9 - 10 1027 123
          XM_017016884.1 XP_016872373.1 9 - 10 7561 123
          XM_017016885.1 XP_016872374.1 8 - 9 7528 123
          XM_017016886.1 XP_016872375.1 8 - 9 7528 123
          XM_017016887.1 XP_016872376.1 9 - 10 1091 123
          XM_017016888.1 XP_016872377.1 9 - 10 1156 123
          XM_017016889.1 XP_016872378.1 8 - 9 1058 123
          XM_017016890.1 XP_016872379.1 8 - 9 7357 123
          XM_017016891.1 XP_016872380.1 8 - 9 1123 123
          XM_017016892.1 XP_016872381.1 8 - 9 1044 123
          XM_017016893.1 XP_016872382.1 7 - 8 7324 123
          XM_017016894.1 XP_016872383.1 7 - 8 7324 123
          XM_017016895.1 XP_016872384.1 9 - 10 1027 123
          XM_017016896.1 XP_016872385.1 8 - 9 995 123
          XM_017016897.1 XP_016872386.1 8 - 9 952 123
          XM_017016898.1 XP_016872387.1 7 - 8 919 123
          XM_017016899.1 XP_016872388.1 7 - 8 919 123
          XM_017016900.1 XP_016872389.1 8 - 9 945 123
          XM_017016901.1 XP_016872390.1 8 - 9 925 123
          XM_017016902.1 XP_016872391.1 8 - 9 944 123
          XM_017016903.1 XP_016872392.1 8 - 9 929 123
          XM_017016904.1 XP_016872393.1 7 - 8 924 123
          XM_017016905.1 XP_016872394.1 7 - 8 910 123
          XM_017016906.1 XP_016872395.1 7 - 8 791 123
          XM_017016907.1 XP_016872396.1 7 - 8 924 123
          XM_017016908.1 XP_016872397.1 7 - 8 913 123
          GenBank mRNA AB082527.1 - 7 - 8 831 123
          AF505653.1 - 7 - 8 869 123
          AK057469.1 - 8 - 9 995 123
          AK294510.1 - 5 - 6 910 123
          AK295031.1 - 7 - 8 954 123
          AK304842.1 - 6 - 7 847 123
          AL133064.1 - 2 - 3 211 123
          BC030555.1 - 8 - 9 1032 5218685 123

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ACBD5

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          macroautophagy
          autophagy of peroxisome
          aggrephagy
          fatty-acyl-CoA binding
          molecular_function
          lipid binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0